missing translation for 'onlineSavingsMsg'
Learn More

Caffeic Acid 3-Beta-D-Glucuronide, TRC

Product Code. 30136920
Change view
Click to view available options
Quantity:
1 mg
2.5 mg
5 mg
Unit Size:
1mg
2.50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30136920 2.5 mg 2.50mg
30006186 1 mg 1mg
30009974 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30136920 Supplier Toronto Research Chemicals Supplier No. TRCC0800152.5MG
 Restricted Item
Estimated Shipment: 13-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1093679-73-2
Molecular Formula C15H16O10
Molecular Weight (g/mol) 356.28
Synonym 5-[(1E)-2-Carboxyethenyl]-2-hydroxyphenyl β-D-Glucopyranosiduronic Acid;
IUPAC Name (2S,3S,4S,5R,6S)-6-[5-[(E)-2-carboxyethenyl]-2-hydroxyphenoxy]-3,4,5-trihydroxyoxane-2-carboxylic acid
SMILES C1=CC(=C(C=C1C=CC(=O)O)OC2C(C(C(C(O2)C(=O)O)O)O)O)O

Specifications

Quantity 2.5 mg
InChI Formula InChI=1S/C15H16O10/c16-7-3-1-6(2-4-9(17)18)5-8(7)24-15-12(21)10(19)11(20)13(25-15)14(22)23/h1-5,10-13,15-16,19-21H,(H,17,18)(H,22,23)/b4-2+/t10-,11-,12+,13-,15+/m0/s1
Formula Weight 356.074
Percent Purity >95%
Chemical Name or Material Caffeic Acid 3-Beta-D-Glucuronide
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.