missing translation for 'onlineSavingsMsg'
Learn More

Cafestol

Product Code. 15416859
Change view
Click to view available options
Quantity:
50 mg
100 mg
Unit Size:
100mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15416859 50 mg 50mg
15406859 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15416859 Supplier Thermo Scientific Alfa Aesar Supplier No. J65355.MF
 Restricted Item
Estimated Shipment: 02-09-2026
Add to basket
This item is not returnable. View return policy

Natural extract from coffee beans which induces glutathione S-transferase | CAS: 469-83-0 | C20H28O3 | 316.44 g/mol

Natural extract from coffee beans which induces glutathione S-transferase

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 469-83-0
Molecular Formula C20H28O3
Molecular Weight (g/mol) 316.44
MDL Number MFCD01075769
InChI Key DNJVYWXIDISQRD-HWUKTEKMSA-N
Synonym cafestol, cafesterol, ccris 1518, unii-ac465t6q6w, 5a,8-methano-5ah-cyclohepta 5,6 naphtho 2,1-b furan-7-methanol, 3b,4,5,6,7,8,9,10,10a,10b,11,12-dodecahydro-7-hydroxy-10b-methyl-, 3bs,5as,7r,8r,10ar,10bs, 5a,8-methano-5ah-cyclohepta 5,6 naphtho 2,1-b furan-7-methanol, 3b,4,5,6,7,8,9,10,10a,10b,11,12-dodecahydro-7-hydroxy-10b-methyl-, 3bs-3balpha,5abeta,7beta,8beta,10aalpha,10bbeta, --cafestol, 1s,4s,12s,13r,16r,17r-17-hydroxymethyl-12-methyl-8-oxapentacyclo 14.2.1.0 1,13 .0 4,12 .0 5,9 nonadeca-5 9 ,6-dien-17-ol, 3bs,5as,7r,8r,10ar,10bs-7-hydroxymethyl-10b-methyl-3b,4,5,6,7,8,9,10,10a,10b,11,12-dodecahydro-5a,8-methanocyclohepta 5,6 naphtho 2,1-b furan-7-ol
PubChem CID 108052
ChEBI CHEBI:3291
IUPAC Name (1S,4S,12S,13R,16R,17R)-17-(hydroxymethyl)-12-methyl-8-oxapentacyclo[14.2.1.0¹,¹³.0⁴,¹².0⁵,⁹]nonadeca-5(9),6-dien-17-ol
SMILES C[C@@]12CCC3=C(C=CO3)[C@H]1CC[C@@]13C[C@@H](CC[C@@H]21)[C@@](O)(CO)C3

Specifications

Melting Point 156°C to 158°C
Color White
Density 1.23
Quantity 50 mg
Merck Index 14,1634
Solubility Information Soluble in dichlormethane, DMSO or ethanol
Formula Weight 316.44
Physical Form Powder
Chemical Name or Material Cafestol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.