Learn More
Specac™ CaF2 Sealed Heatable Liquid Flow Cell VTCH Incompatible
Description
This cell is designed for use in Specac heating jackets GS20710 and GS20730. This is a sealed heatable liquid flow cell with CaF2 windows and choice of pathlength from 0.025mm up to 1.0mm. This cell is fitted with 1/16in. Swagelok fittings for flow purposes.
Specifications
Specifications
| Type | CaF2 Sealed Heatable Liquid Flow Cell |
| Description | Pathlength from 0.025 mm up to 1.0 mm |