Learn More
Specac™ CaF2 Sealed Heatable Liquid Flow Cell VTCH Compatible
Product Code. 15270218
Encompass_Preferred
Click to view available options
Type:
CaF2 Sealed Heatable Liquid Flow Cell
Product Code. 15270218
Brand: Specac™ GS20562
This item requires a price update prior to being ordered.
Request a Price for this item here.
This cell is designed for use in the Specac variable temperature cell GS21525. This is a sealed heatable liquid flow cell with CaF2 windows and choice of pathlength from 0.025mm up to 1.0mm. This cell is fitted with 1/16in. Swagelok fittings for flow purposes.
Specifications
Type CaF2 Sealed Heatable Liquid Flow Cell
Description Pathlength from 0.025 mm up to 1.0 mm