missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Positive Samples: HeLa, K-562 Immunogen sequence: MLEELECGAP GARGAATAMD CKDRPAFPVK KLIQARLPFK RLNLVPKGKA DDMSDDQGTS VQSKSPDLEA SLDTLENNCH VGSDIDFRPK LVNGKGPLDN FLRNRIETSI GQSTVIIDLT EDSNEQPDSL VDHNKLNSEA SPSREAINGQ REDTGDQQGL LKAIQNDKLA FPGETLSDIP CKTEEEGVGC GGAGRRGDSQ.
CAF1 p150 is part of the chromatin assembly factor 1 (CAF-1) complex, a key player in DNA repair and replication processes. CAF-1, composed of p150 (CHAF1A), p60 (CHAF1B), and p48 subunits, functions primarily in the deposition of newly synthesized histones H3 and H4 onto nascent DNA strands, ensuring chromatin is properly assembled during DNA replication and after repair. The p150 subunit is the largest of the CAF-1 complex and is critical for binding DNA and recruiting CAF-1 to replication sites, facilitating histone deposition in a process vital for maintaining genome stability and epigenetic inheritance. Beyond just chromatin formation, CAF1 p150 is involved in influencing gene silencing and the maintenance of heterochromatin, impacting gene regulation notably during development and in response to DNA damage. Its role has implications in oncogenesis, where dysregulation of CAF1 p150 can lead to altered chromatin structures and contribute to tumorigenesis, marking it as an important target in cancer research.
Specifications
Specifications
| Antigen | CAF1 p150 |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Concentration | 0.98 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 |
| Gene | CHAF1A |
| Gene Accession No. | Q13111 |
| Gene Alias | AL023013; AL024058; Cac1p; CAF; CAF1; CAF-1; CAF-1 subunit A; CAF1B; CAF1P150; CAF-I 150 kDa subunit; CAF-I p150; CAF-Ip150; Caip150; CHAF1A; Chromatin assembly factor 1 subunit A; chromatin assembly factor 1, subunit A (p150); chromatin assembly factor I (150 kDa); chromatin assembly factor I p150 subunit; CTB-50L17.7; hp150; MGC71229; P150 |
| Gene Symbols | CHAF1A |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?
