missing translation for 'onlineSavingsMsg'
Learn More

Caesium Chloride, 99.5+%, for Ultracentrifuge Work, Fisher Chemical

Product Code. 10395070 Shop All Fisher Chemical Products
Change view
Click to view available options
100g, Plastic powder jar
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
10395070 100g, Plastic powder jar
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10395070 Supplier Fisher Chemical Supplier No. C/0682/48
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

Boiling Point 1290°C
Melting Point 646°C
CAS 7647-17-8
Color White
pH 5 to 9
Physical Form Powder/Solid
Quantity 100 g
Molecular Formula ClCs
MDL Number 10955
Formula Weight 168.36g/mol
Percent Purity 99.5%
Packaging Plastic powder jar
Show More Show Less
compliance-icons
Product Identifier
  • Cesium chloride
Signal Word
  • Warning
Hazard Category
  • Reproductive toxicity Category 2
Hazard Statement
  • H361-Suspected of damaging fertility or the unborn child.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.