missing translation for 'onlineSavingsMsg'
Learn More

Caesium Chloride, 99.5+%, Certified AR for Analysis, Fisher Chemical™

Product Code. 10588030 Shop All Fisher Chemical Products
Click to view available options
Quantity:
25 g
100 g
250 g
1 kg
Unit Size:
100g
1kg
250g
25g
This item is not returnable. View return policy

Product Code. 10588030

Brand: Fisher Chemical C/0680/60

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

ClCs, CAS Number-7647-17-8, 1kg, 1290 deg.C, White, 168.36g/mol, 646 deg.C, 10955, Plastic powder jar, 5 to 9, Powder Solid

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7647-17-8
Molecular Formula ClCs
Molecular Weight (g/mol) 168.36
MDL Number MFCD00010955
InChI Key AIYUHDOJVYHVIT-UHFFFAOYSA-M
IUPAC Name caesium(1+) chloride
SMILES [Cl-].[Cs+]

Specifications

Boiling Point 1290°C
Melting Point 646°C
Color White
pH 5 to 9
Physical Form Powder
Quantity 1 kg
Formula Weight 168.36g/mol
Percent Purity 99.5%
Packaging Plastic powder jar
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.