missing translation for 'onlineSavingsMsg'
Learn More

Caerulomycin A, MedChemExpress

Product Code. 16471209
Encompass_Preferred
Click to view available options
Quantity:
1 mg
5 mg
10 mg
Unit Size:
10mg
1mg
5mg
This item is not returnable. View return policy

Product Code. 16471209

Brand: Medchem Express HY114495

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Caerulomycin A (Cerulomycin; Caerulomycin), an antifungal compound, induces generation of T cells, enhances TGF-β-Smad3 protein signaling via suppressing interferon-γ-induced STAT1 signaling. Antifungal and antibiotic activity, and used in autoimmune diseases.

Caerulomycin A (Cerulomycin

Specifications

Chemical Name or Material Caerulomycin A
Physical Form Solid
CAS 21802-37-9
Color Off-White
Quantity 1 mg
Purity Grade Notes Research
Molecular Formula C12H11N3O2
For Use With (Application) COVID-19-immunoregulation
Synonym Cerulomycin Caerulomycin
Solubility Information DMSO : 150 mg/mL (654.36 mM; Need ultrasonic)
SMILES COC1=CC(C2=NC=CC=C2)=NC(/C=N/O)=C1.[(E)]
Molecular Weight (g/mol) 229.23
Formula Weight 229.23
Percent Purity 98.0%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.