missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CADPS2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5109477
This item is not returnable.
View return policy
Description
Immunogen sequence: MLDPSSSEEE SDEGLEEESR DVLVAAGSSQ RAPPAPTREG RRDAPGRAGG GGAARSVSPS PSVLSEGRDE PQRQLDDEQE.
CAPS2 and the related protein CAPS1 encode novel neural/endocrine-specific cytosolic and peripheral membrane proteins. Both are essential components of the synaptic vesicle priming machinery and are required for the Ca2+-regulated exocytosis of secretory vesicles; CAPS-deficient neurons contain no or very few fusion competent synaptic vesicles, causing a selective impairment of fast phasic transmitter release. Like CAPS1, numerous isoforms of CAPS2 are known to exist. These isoforms show distinct expression patterns in the brain. For example, CAPS2b showed high expression in the developing cerebellum. Furthermore, one version of CAPS2 mRNA that lacks exon 3 is detected in some autistic patients, suggesting that the differential expression pattern of CAPS2 is involved in neuronal development.
Specifications
| CADPS2 | |
| Polyclonal | |
| Unconjugated | |
| CADPS2 | |
| A230044C21Rik; Ca++-dependent secretion activator 2; Ca2+ dependent secretion activator 2; Ca2+-dependent activator protein for secretion 2; Cadps2; calcium dependent secretion activator 2; calcium-dependent activator protein for secretion 2; Calcium-dependent secretion activator 2; calcyphosin-2; calcyphosine 2; Calcyphosine-2; calcyphosphine 2; Caps2; CAPS-2; cerebellum postnatal development associated protein 2; Cpd2; KIAA1591; RGD1559440; UG0636c06 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 312166, 320405, 93664 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 0.86 mg/mL | |
| PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
| Q86UW7, Q8BYR5 | |
| CADPS2 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-80 of human CADPS2 (NP_001009571.2). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction