missing translation for 'onlineSavingsMsg'
Learn More

Cadmium wire, 2.0mm (0.08in) dia, Puratronic™, 99.998% (metals basis), Thermo Scientific Chemicals

Catalog Number 11366368
Click to view available options
Quantity:
1 m
25 cm
This item is not returnable. View return policy
7440-43-9
Cd
112.41
MFCD00010914
BDOSMKKIYDKNTQ-UHFFFAOYSA-N
kadmium, compounds, colloidal, cadmium, elemental, kadmium german, unii-00bh33gngh, ccris 112, hsdb 282, 00bh33gngh, cadmium and compounds
23973
CHEBI:22977
cadmium
[Cd]
This item is not returnable. View return policy
7440-43-9
Cd
112.41
MFCD00010914
BDOSMKKIYDKNTQ-UHFFFAOYSA-N
kadmium, compounds, colloidal, cadmium, elemental, kadmium german, unii-00bh33gngh, ccris 112, hsdb 282, 00bh33gngh, cadmium and compounds
23973
CHEBI:22977
cadmium
[Cd]

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Quantity 1 m
Percent Purity 99.998%
Grade Puratronic™
Weight ≈27.2g/m
Form Wire
Assay (metals basis)
Chemical Name or Material Cadmium wire, 2.0mm (0.08 in.) dia.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'