missing translation for 'onlineSavingsMsg'
Learn More

Cadmium tellurite, 99%

Product Code. 11385089
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11385089 25 g 25g
11375089 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11385089 Supplier Thermo Scientific Alfa Aesar Supplier No. A17476.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 15851-44-2 | CdO3Te | 288.01 g/mol

It is employed in the preparation, characterization and structure of Ca8[AlO2]12Te2 and Cd8[AlO2]12Te2; two new members of the sodalite family.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is employed in the preparation, characterization and structure of Ca8[AlO2]12Te2 and Cd8[AlO2]12Te2; two new members of the sodalite family.

Solubility
Insoluble in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 15851-44-2
Molecular Formula CdO3Te
Molecular Weight (g/mol) 288.01
MDL Number MFCD00049413
InChI Key GKMPTXZNGKKTDU-UHFFFAOYSA-L
Synonym cadmium tellurite, cadmium 2+ tellurite, cadmium tellurium trioxide, cd.o3te, acmc-20alj5, cadmium 2+ ion tellurite, telluric acid h2teo3 , cadmium salt 1:1, tellurous acid cadmium salt, telluric acid h2teo3 ,cadmium salt 1:1
PubChem CID 167475
IUPAC Name cadmium(2+);tellurite
SMILES [Cd++].[O-][Te]([O-])=O

Specifications

Quantity 25 g
UN Number UN2570
Solubility Information Insoluble in water.
Formula Weight 288.01
Percent Purity 99%
Chemical Name or Material Cadmium tellurite

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.