missing translation for 'onlineSavingsMsg'
Learn More

Cadmium sulfide, 99.999% (metals basis)

Product Code. 15462267
Change view
Click to view available options
Quantity:
50 g
250 g
Unit Size:
250g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15462267 250 g 250g
15452267 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15462267 Supplier Thermo Scientific Alfa Aesar Supplier No. 045455.30
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Used as pigment in the chemical industry; photoresistors; optical-electronic pairs, photodiodes, solar batteries and phosphors. Single crystals of this compound are used for the preparation of optical devices operating in the infrared regaion of spectrum. | CAS: 1306-23-6 | CdS | 144.47 g/mol

Cadmium sulfide is used as pigment in the chemical industry; photoresistors; optical-electronic pairs, photodiodes, solar batteries and phosphors. Single crystals of this compound are used for the preparation of optical devices operating in the infrared regaion of spectrum. It is a direct band gap semiconductor with a band gap band from 2.42 to 2.57 eV.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1306-23-6
Molecular Formula CdS
Molecular Weight (g/mol) 144.47
MDL Number MFCD00010922
InChI Key FRLJSGOEGLARCA-UHFFFAOYSA-N
Synonym cadmium sulfide, cadmium sulphide, capsebon, orange cadmium, cadmopur yellow, jaune brilliant, aurora yellow, cadmium orange, cadmium yellow, ferro yellow
PubChem CID 14783
ChEBI CHEBI:50833
IUPAC Name sulfanylidenecadmium
SMILES [S--].[Cd++]

Specifications

Melting Point 980°C (sublimation)
Physical Form Pieces
Quantity 250 g
UN Number UN3077
Solubility Information Insoluble in water.
Formula Weight 144.46
Percent Purity 99.999%
Packaging Plastic bottle
Specific Gravity 4.82
Density 4.82
Chemical Name or Material Cadmium sulfide
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.