missing translation for 'onlineSavingsMsg'
Learn More

Cadmium sulfate octahydrate, ACS, 98.0-102.0%

Product Code. 11361709
Change view
Click to view available options
Quantity:
25 g
250 g
1 kg
Unit Size:
1kg
250g
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11361709 1 kg 1kg
11381709 25 g 25g
11371709 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11361709 Supplier Thermo Scientific Alfa Aesar Supplier No. 012340.A1
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Used in Standard Weston Cell, as electrolytes in electroplating, in phosphors, in manufacture of standard cadmium elements, and as a catalyst in Marsh test for AS | CAS: 7790-84-3 | 3CdSO4·8H2O

Cadmium sulfate mainly used in electroplating of cadmium, which is used in electronic circuits. It is used as a precursor for cadmium based pigments such as cadmium sulfide and pharmaceutical intermediates. It is used in the standard weston cell.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7790-84-3
MDL Number MFCD00151655

Specifications

Melting Point >40°C to H2O
Color White
Physical Form Crystalline
Quantity 1 kg
Assay Percent Range 98.0 to 102.0%
Molecular Formula 3CdSO4·8H2O
Solubility Information Highly soluble in water. Partially soluble in methanol and ethyl acetate. Insoluble in ethanol.
Formula Weight 769.50 (208.46 Anhydrous)
Grade ACS
Refractive Index 1.565
Appearance White crystalline
Density 3.08
Chemical Name or Material Cadmium sulfate octahydrate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.