missing translation for 'onlineSavingsMsg'
Learn More

Cadmium sulfate hydrate, 98%

Product Code. 11489452
Click to view available options
Quantity:
100 g
500 g
2500 g
Unit Size:
100g
2500g
500g
This item is not returnable. View return policy

Product Code. 11489452

Brand: Thermo Scientific Alfa Aesar A10168.36

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 15244-35-6 | CdO4S | 208.47 g/mol

Cadmium sulfate hydrate is used for formulating screens or for optimization. It is also used as pharmaceutical intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Cadmium sulfate hydrate is used for formulating screens or for optimization. It is also used as pharmaceutical intermediates.

Solubility
Soluble in water (1130g/L ).

Notes
Store in cool, dry conditions in well sealed containers. keep container tightly closed. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 15244-35-6
Molecular Formula CdO4S
Molecular Weight (g/mol) 208.47
MDL Number MFCD00151655
InChI Key QCUOBSQYDGUHHT-UHFFFAOYSA-L
IUPAC Name cadmium(2+) sulfate
SMILES [Cd++].[O-]S([O-])(=O)=O

Specifications

Melting Point 41.5°C (trans pt)
Quantity 500 g
UN Number UN2570
Merck Index 14,1627
Solubility Information Soluble in water (1130g/L ).
Formula Weight 208.46 (Anhydrous)
Percent Purity 98%
Odor Odorless
Refractive Index 1.565
Density 3.08
Chemical Name or Material Cadmium sulfate hydrate
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.