missing translation for 'onlineSavingsMsg'
Learn More

Cadmium Sulfate, Extra Pure, SLR, Fisher Chemical™

Product Code. 10754941 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10754941 100 g 100g
10610331 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10754941 Supplier Fisher Chemical Supplier No. C/0480/48

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 7790-84-3
Molecular Formula CdH2O5S
Molecular Weight (g/mol) 226.49
MDL Number 2253000
InChI Key VRFRVKRTIMIRNG-UHFFFAOYSA-L
IUPAC Name cadmium(2+) hydrate sulfate
SMILES O.[Cd++].[O-]S([O-])(=O)=O

Specifications

Melting Point 41°C
Color White
pH 4.5 to 6.0
Physical Form Solid
Quantity 100 g
Formula Weight 769.51g/mol
Packaging Plastic powder jar
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Cadmium sulfate octahydrate
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Acute toxicity Category 2
  • Germ cell mutagenicity Category 1B
  • Carcinogenicity Category 1B
  • Reproductive toxicity Category 1B
  • ACUTE AQUATIC Acute 1
  • LONG-TERM AQUATIC HAZARD Chronic 1
Hazard Statement
  • H350-May cause cancer.
  • H340-May cause genetic defects.
  • H360FD-May damage fertility. May damage the unborn child.
  • H330-Fatal if inhaled.
  • H301-Toxic if swallowed.
  • H372-Causes damage to organs.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P310-Immediately call a POISON CENTER/doctor/.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P273-Avoid release to the environment.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.