missing translation for 'onlineSavingsMsg'
Learn More

Cadmium sulfate, ACS reagent, anhydrous

Product Code. p-4813375 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Glass bottle
Unit Size:
100g
2.5kg
500g
This item is not returnable. View return policy

Product Code. 12615107

Brand: Thermo Scientific Acros 447751000

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 10124-36-4 | CdO4S | 208.46 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10124-36-4
Molecular Formula CdO4S
Molecular Weight (g/mol) 208.46
InChI Key QCUOBSQYDGUHHT-UHFFFAOYSA-L
Synonym cadmium sulfate, cadmium sulphate, cadmium monosulfate, sulfuric acid, cadmium salt 1:1, cadmium sulfuricum, cadmium sulfate 1:1, cadmium sulfate, tetrahydrate, cadmium 2+ sulfate, caswell no. 136c, unii-947unf3z6o
PubChem CID 24962
ChEBI CHEBI:50292
IUPAC Name cadmium(2+);sulfate
SMILES [O-]S(=O)(=O)[O-].[Cd+2]

Specifications

Melting Point 1000.0°C
Quantity 100 g
Linear Formula CdSO4
Merck Index 15,1632
Solubility Information Solubility in water: 1130g/L (20°C). Other solubilities: insoluble in alcohol, acids, acetone and ammonia
Formula Weight 208.46
Grade ACS Reagent
Packaging Glass bottle
Chemical Name or Material Cadmium sulfate
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Cadmium sulphate
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 2
  • Acute toxicity Category 3
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Carcinogenicity Category 1B
  • Germ cell mutagenicity Category 1B
  • Reproductive toxicity Category 1B
  • Specific target organ toxicity Category 1
Hazard Statement
  • H301-Toxic if swallowed.
  • H330-Fatal if inhaled.
  • H340-May cause genetic defects.
  • H350-May cause cancer.
  • H360FD-May damage fertility. May damage the unborn child.
  • H372-Causes damage to organs.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contains : Cadmium sulphate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.