missing translation for 'onlineSavingsMsg'
Learn More

Cadmium Sulfate, ACS Reagent, ≥99%, Solstice

Product Code. 15625300 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Poly Bottle
Unit Size:
100g
2.5kg
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
15625300 500 g Poly Bottle 500g
15605300 100 g Poly Bottle 100g
15615300 2.5 kg Poly Bottle 2.5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15625300 Supplier Honeywell-Fluka Supplier No. 383082500G
  • ONLINE EXCLUSIVE PRICE
    £311.00
    £285.95 / 500g
    Save £25.05
    8% Off
Request Bulk or Custom Formats

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

Currently Unavailable: We have no information when this product will be available.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

ACS Reagent,≥99.0%

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10124-36-4
Molecular Formula CdO4S
Molecular Weight (g/mol) 208.47
MDL Number MFCD00010923
InChI Key QCUOBSQYDGUHHT-UHFFFAOYSA-L
Synonym cadmium sulfate, cadmium sulphate, cadmium monosulfate, sulfuric acid, cadmium salt 1:1, cadmium sulfuricum, cadmium sulfate 1:1, cadmium sulfate, tetrahydrate, cadmium 2+ sulfate, caswell no. 136c, unii-947unf3z6o
PubChem CID 24962
ChEBI CHEBI:50292
IUPAC Name cadmium(2+);sulfate
SMILES [O-]S(=O)(=O)[O-].[Cd+2]

Specifications

Melting Point 1000°C
Color Colorless
Quantity 500 g
Loss on Drying ≤1.0%
Packaging Poly Bottle
Appearance Crystalline
Density 4.69g/cm3
Linear Formula CdSO4
UN Number UN2570
Formula Weight 208.47g/mol
Percent Purity ≥99%
Chemical Name or Material Cadmium sulfate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.