missing translation for 'onlineSavingsMsg'
Learn More

Cadmium Sulfate 8/3-Hydrate, puriss. p.a. ACS Reagent, ≥99.0%, Solstice

Product Code. 15681890 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Poly Bottle
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
15681890 25 g Poly Bottle 25g
15671890 100 g Poly Bottle 100g
15691890 500 g Poly Bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15681890 Supplier Honeywell-Fluka Supplier No. 2092025G
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Puriss. p.a., ACS Reagent,≥99.0% (calc. based on CdSO4 · 8/3H2O, KT)

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7790-84-3
MDL Number MFCD02253000

Specifications

Melting Point 41.5°C
Color Colorless
Physical Form Solid
Quantity 25 g
Linear Formula CdSO4 · 8/3H2O
UN Number UN2570
Formula Weight 256.57g/mol
Percent Purity ≥99.0% (calc. based on CdSO4.83H2O, KT)
Grade ACS/puriss. p.a.
Packaging Poly Bottle
Density 3.09g/cu.cm
Chemical Name or Material Cadmium Sulfate 8/3-Hydrate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.