missing translation for 'onlineSavingsMsg'
Learn More

Cadmium Sulfate 8/3-Hydrate, Honeywell Fluka™

Product Code. p-7071769 Shop All Honeywell Fluka Products
Click to view available options
Quantity:
500 g
1 kg
Unit Size:
1kg
500g
This item is not returnable. View return policy

Product Code. 15651070

Brand: Honeywell-Fluka 11720500G

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Puriss.,≥98% (complexometric)

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7790-84-3
MDL Number MFCD02253000

Specifications

Quantity 500 g
Linear Formula CdSO4 · 8/3H2O
UN Number 2570
Formula Weight 256.57g/mol
Percent Purity ≥98% (complexometric)
Grade puriss.
Chemical Name or Material Cadmium Sulfate 8/3-Hydrate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.