missing translation for 'onlineSavingsMsg'
Learn More

Cadmium Sulfate 8/3-Hydrate, Honeywell Fluka™

Product Code. 15651070 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
500 g
1 kg
Unit Size:
1kg
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15651070 500 g 500g
15641070 1 kg 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15651070 Supplier Honeywell-Fluka Supplier No. 11720500G

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Puriss.,≥98% (complexometric)

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7790-84-3
MDL Number MFCD02253000

Specifications

Quantity 500 g
Linear Formula CdSO4 · 8/3H2O
UN Number 2570
Formula Weight 256.57g/mol
Percent Purity ≥98% (complexometric)
Grade puriss.
Chemical Name or Material Cadmium Sulfate 8/3-Hydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.