missing translation for 'onlineSavingsMsg'
Learn More

Cadmium standard solution, for AAS, 1mg/mL Cd in 0.5N HNO3, Thermo Scientific Chemicals

£21.20 - £72.20

Specifications

Chemical Name or Material Cadmium standard solution
Concentration 997 to 1003 μg/mL of Cd(2+) in 0.5N HNO3 (20°C)
Name Note 1 mg/ml Cd in 0.5N HNO3
CAS 10022-68-1, 7697-37-2, 7732-18-5
Packaging Plastic bottle
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10501071
View Documents Promotion Details
Thermo Scientific Acros
195871000
100 mL Plastic bottle
£21.20
100mL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
10782811
View Documents Promotion Details
Thermo Scientific Acros
195875000
500 mL Plastic bottle
£72.20
500mL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

for atomic adsorption spectroscopy
Specifications

Specifications

Cadmium standard solution
1 mg/ml Cd in 0.5N HNO3
Plastic bottle
MFCD00149626
IZIPRBSCGMISKX-UHFFFAOYSA-N
cadmium(2+) bis(nitric acid)
56924536
308.48
1.0120g/mL
Soluble in water
997 to 1003 μg/mL of Cd(2+) in 0.5N HNO3 (20°C)
10022-68-1, 7697-37-2, 7732-18-5
CdH2N2O6
15, 1627
[Cd++].O[N+]([O-])=O.O[N+]([O-])=O
238.44
CHEBI:86156
AAS
1.012
Solution
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
May be corrosive to metals.
Harmful to aquatic life with long lasting effects.
May cause cancer.

GHS P Statement:
Obtain special instructions before use.
Use personal protective equipment as required.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Avoid release to the environment.
Avoid breathing dust/fume/gas/mist/vapors/spray.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

Product Identifier
  • Cadmium atomic absorption standard solution

Signal Word
  • Danger

Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Carcinogenicity Category 1A
  • Serious eye damage/eye irritation Category 1
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 2

Hazard Statement
  • H318-Causes serious eye damage.
  • H350-May cause cancer.
  • H315-Causes skin irritation.
  • H412-Harmful to aquatic life with long lasting effects.
  • H290-May be corrosive to metals.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only