missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cadmium selenide, 99.995% (metals basis), Thermo Scientific Chemicals
35.80 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
5 g
25 g
100 g
Description
It ia a photoconductive and semiconductor material used in a cell where a fast response time and high sensitivity to longer wavelengths of light is required; also used as a red pigment.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | >1350°C |
Color | Black to Brown-Red |
Physical Form | Sintered Lump |
Quantity | 5 g |
UN Number | UN3077 |
Merck Index | 14,1626 |
Solubility Information | Insoluble in water but soluble in acids. |
Formula Weight | 191.36 |
Percent Purity | 99.995% |
Odor | Odorless |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'