missing translation for 'onlineSavingsMsg'
Learn More

Cadmium selenide, 99.995% (metals basis), Thermo Scientific Chemicals

Product Code. 11351708
Change view
Click to view available options
Quantity:
2 g
10 g
50 g
Packaging:
Plastic bottle
Plastic vial
Unit Size:
10g
2g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
11351708 10 g Plastic bottle 10g
11331708 2 g Plastic vial 2g
11341708 50 g Plastic bottle 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11351708 Supplier Thermo Scientific Alfa Aesar Supplier No. 011057.09
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Used as a n-type semiconductor and its nanoparticles are used in laser diodes, opto-electronic devices, nanosensing, biomedical imaging and high efficiency solar cells. Since, it is transparent to infrared light, it finds application in photoresistors and windows for instruments utilizing IR light.

Cadmium selenide is used as a n-type semiconductor and its nanoparticles are used in laser diodes, opto-electronic devices, nanosensing, biomedical imaging and high efficiency solar cells. Since, it is transparent to infrared light, it finds application in photoresistors and windows for instruments utilizing IR light.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1306-24-7
Molecular Formula CdSe
Molecular Weight (g/mol) 191.39
MDL Number MFCD00010917
InChI Key AQCDIIAORKRFCD-UHFFFAOYSA-N
Synonym cadmium selenide, cadmium ii selenide, cadmium 2+ selenide, cadmium selenide nanotubes, cadmium selenide trace metals basis 1g, cadmium selenide candot quantum dot core , 50umol/l in hexane, lumidot tm cdse, 480, core-type quantum dots, 5 mg/ml in toluene, lumidot tm cdse, 520, core-type quantum dots, 5 mg/ml in toluene
PubChem CID 14784
ChEBI CHEBI:50834
IUPAC Name selanylidenecadmium
SMILES [Se]=[Cd]

Specifications

Melting Point >1350°C
Physical Form Powder
Quantity 10 g
UN Number UN3077
Formula Weight 191.36
Percent Purity 99.995%
Packaging Plastic bottle
Specific Gravity 5.81
Chemical Name or Material Cadmium selenide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.