missing translation for 'onlineSavingsMsg'
Learn More

Cadmium rod, 12.7mm (0.5in) dia x 23cm (9.0in), Puratronic™, 99.999+% (metals basis), Thermo Scientific Chemicals

Catalog Number 11355739
Click to view available options
Quantity:
1 Pc.
5 Pc.
This item is not returnable. View return policy
7440-43-9
Cd
112.41
MFCD00010914
BDOSMKKIYDKNTQ-UHFFFAOYSA-N
kadmium, compounds, colloidal, cadmium, elemental, kadmium german, unii-00bh33gngh, ccris 112, hsdb 282, 00bh33gngh, cadmium and compounds
23973
CHEBI:22977
cadmium
[Cd]
This item is not returnable. View return policy
7440-43-9
Cd
112.41
MFCD00010914
BDOSMKKIYDKNTQ-UHFFFAOYSA-N
kadmium, compounds, colloidal, cadmium, elemental, kadmium german, unii-00bh33gngh, ccris 112, hsdb 282, 00bh33gngh, cadmium and compounds
23973
CHEBI:22977
cadmium
[Cd]

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Quantity 1 Pc.
Percent Purity ≥99.999%
Grade Puratronic™
Weight ≈250g/rod
Form Rod
Assay (metals basis)
Chemical Name or Material Cadmium rod, 12.7mm (0.5 in.) dia. x 23cm (9.0 in.)
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Cadmium rod
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 2
  • Germ cell mutagenicity Category 2
  • Carcinogenicity Category 1B
  • Reproductive toxicity Category 2
  • Specific target organ toxicity Category 1
  • ACUTE AQUATIC Acute 1
  • LONG-TERM AQUATIC HAZARD Chronic 1
Hazard Statement
  • H330-Fatal if inhaled.
  • H341-Suspected of causing genetic defects.
  • H350-May cause cancer.
  • H361fd-Suspected of damaging fertility. Suspected of damaging the unborn child.
  • H372-Causes damage to organs.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P284-¢In case of inadequate ventilation! wear respiratory protection.
  • P201-Obtain special instructions before use.
  • P320-Specific treatment is urgent (see on this label).
  • P405-Store locked up.
  • P501-Dispose of contents/container to .

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'