missing translation for 'onlineSavingsMsg'
Learn More

Cadmium rod, 12.7mm (0.5in) dia x 23cm (9.0in), Puratronic™, 99.999+% (metals basis), Thermo Scientific Chemicals

£215.00 - £952.00

Chemical Identifiers

CAS 7440-43-9
Molecular Formula Cd
Molecular Weight (g/mol) 112.41
MDL Number MFCD00010914
InChI Key BDOSMKKIYDKNTQ-UHFFFAOYSA-N
Synonym kadmium, compounds, colloidal, cadmium, elemental, kadmium german, unii-00bh33gngh, ccris 112, hsdb 282, 00bh33gngh, cadmium and compounds
PubChem CID 23973
ChEBI CHEBI:22977
IUPAC Name cadmium
SMILES [Cd]
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11355739
View Documents Promotion Details
Thermo Scientific Alfa Aesar
010120.KG
1 Pc.
£215.00
Each
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
15424735
View Documents Promotion Details
Thermo Scientific Alfa Aesar
010120.KJ
5 Pc.
£952.00
Pack of 5
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

7440-43-9
112.41
BDOSMKKIYDKNTQ-UHFFFAOYSA-N
23973
cadmium
Cd
MFCD00010914
kadmium, compounds, colloidal, cadmium, elemental, kadmium german, unii-00bh33gngh, ccris 112, hsdb 282, 00bh33gngh, cadmium and compounds
CHEBI:22977
[Cd]

Specifications

7440-43-9
MFCD00010914
BDOSMKKIYDKNTQ-UHFFFAOYSA-N
cadmium
23973
≥99.999%
≈250g/rod
(metals basis)
Cd
kadmium, compounds, colloidal, cadmium, elemental, kadmium german, unii-00bh33gngh, ccris 112, hsdb 282, 00bh33gngh, cadmium and compounds
[Cd]
112.41
CHEBI:22977
Puratronic™
Rod
Cadmium rod, 12.7mm (0.5 in.) dia. x 23cm (9.0 in.)
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H330-H350-H372-H341-H361
Fatal if inhaled.
May cause cancer.
Causes damage to organs through prolonged or repeated exposure.
Suspected of causing genetic defects.
Suspected of damaging fertility or the unborn child.

P201-P202-P260-P264b-P270-P271-P281-P284-P304+P340-P308+P313-P310-P501c

H330-H341-H350-H361fd-H372-H500

missing translation for 'einecsNumber' : 231-152-8

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

Product Identifier
  • Cadmium rod

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 2
  • Germ cell mutagenicity Category 2
  • Carcinogenicity Category 1B
  • Reproductive toxicity Category 2
  • Specific target organ toxicity Category 1
  • ACUTE AQUATIC Acute 1
  • LONG-TERM AQUATIC HAZARD Chronic 1

Hazard Statement
  • H330-Fatal if inhaled.
  • H341-Suspected of causing genetic defects.
  • H350-May cause cancer.
  • H361fd-Suspected of damaging fertility. Suspected of damaging the unborn child.
  • H372-Causes damage to organs.
  • H410-Very toxic to aquatic life with long lasting effects.

Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P284-¢In case of inadequate ventilation! wear respiratory protection.
  • P201-Obtain special instructions before use.
  • P320-Specific treatment is urgent (see on this label).
  • P405-Store locked up.
  • P501-Dispose of contents/container to .

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only