missing translation for 'onlineSavingsMsg'
Learn More

Cadmium rod, 12.7mm (0.5in) dia, 99.99+% (metals basis), Thermo Scientific Chemicals

Product Code. 11321168
Click to view available options
Quantity:
12 cm
24 cm
5 x 24 cm
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7440-43-9
Molecular Formula Cd
Molecular Weight (g/mol) 112.41
MDL Number MFCD00010914
InChI Key BDOSMKKIYDKNTQ-UHFFFAOYSA-N
Synonym kadmium, compounds, colloidal, cadmium, elemental, kadmium german, unii-00bh33gngh, ccris 112, hsdb 282, 00bh33gngh, cadmium and compounds
PubChem CID 23973
ChEBI CHEBI:22977
IUPAC Name cadmium
SMILES [Cd]

Specifications

Quantity 24 cm
Percent Purity ≥99.99%
Weight ≈11.0g/cm
Form Rod
Assay (metals basis)
Chemical Name or Material Cadmium rod, 12.7mm (0.5 in.) dia.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'