missing translation for 'onlineSavingsMsg'
Learn More

Cadmium perchlorate hexahydrate

Product Code. 11332028
Change view
Click to view available options
Quantity:
50 g
250 g
Unit Size:
250g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11332028 250 g 250g
11342028 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11332028 Supplier Thermo Scientific Alfa Aesar Supplier No. 012936.30
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 10326-28-0 | CdCl2H12O14 | 419.396 g/mol

It is employed as intermediate in chemical synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10326-28-0
Molecular Formula CdCl2H12O14
Molecular Weight (g/mol) 419.396
MDL Number MFCD00015992
InChI Key PQJZOTJHMRJOEM-UHFFFAOYSA-L
Synonym cadmium perchlorate hexahydrate, acmc-1c6hb, cd.2clo4.6h2o, cadmium ii perchlorate hexahydrate
PubChem CID 71317310
IUPAC Name cadmium(2+);diperchlorate;hexahydrate
SMILES O.O.O.O.O.O.[O-]Cl(=O)(=O)=O.[O-]Cl(=O)(=O)=O.[Cd+2]

Specifications

Color White
Physical Form Crystals
Quantity 250 g
Solubility Information Insoluble in water.
Formula Weight 419.39 (311.30 Anhydrous)
Sensitivity Hygroscopic
Chemical Name or Material Cadmium perchlorate hexahydrate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.