missing translation for 'onlineSavingsMsg'
Learn More

Cadmium oxide, 99%, pure

Product Code. 15299716 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
250 g
1 kg
Unit Size:
1kg
250g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15299716 250 g 250g
15279716 1 kg 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15299716 Supplier Thermo Scientific Acros Supplier No. 223792500
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1306-19-0 | CdO | 128.41 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1306-19-0
Molecular Formula CdO
Molecular Weight (g/mol) 128.41
MDL Number MFCD00010921
InChI Key CFEAAQFZALKQPA-UHFFFAOYSA-N
Synonym cadmium oxide, cadmium monoxide, cadmium fume, aska-rid, kadmu tlenek polish, caswell no. 136aa, ccris 115, epa pesticide chemical code 236200, dsstox_cid_4715
PubChem CID 14782
IUPAC Name oxocadmium
SMILES [O--].[Cd++]

Specifications

Boiling Point 1385.0°C
Quantity 250 g
Assay Percent Range 98.5% min. (Complexometry)
Linear Formula CdO
Merck Index 15,1628
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in diluted acids and alkalies
Formula Weight 128.41
Percent Purity 99%
Grade Pure
Chemical Name or Material Cadmium oxide
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Cadmium oxide, brown
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 2
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Carcinogenicity Category 1B
  • Germ cell mutagenicity Category 2
  • Reproductive toxicity Category 2
  • Specific target organ toxicity Category 1
Hazard Statement
  • H372-Causes damage to organs.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
  • H350-May cause cancer.
  • H341-Suspected of causing genetic defects.
  • H361fd-Suspected of damaging fertility. Suspected of damaging the unborn child.
  • H330-Fatal if inhaled.
Precautionary Statement
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.