missing translation for 'onlineSavingsMsg'
Learn More

Cadmium nitrate tetrahydrate, Puratronic™, 99.999% (metals basis)

Product Code. p-4407300
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
This item is not returnable. View return policy

Product Code. 11340889

Brand: Thermo Scientific Alfa Aesar 010663.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 10022-68-1 | CdH2N2O6 | 238.44 g/mol

It is used for coloring glass and porcelain, and as a flash powder in photography. It is used as a laboratory reagent, in preparation of cadmium salts.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10022-68-1
Molecular Formula CdH2N2O6
Molecular Weight (g/mol) 238.44
MDL Number MFCD00149626
InChI Key IZIPRBSCGMISKX-UHFFFAOYSA-N
Synonym cadmium nitrate tetrahydrate, nitric acid, cadmium salt, tetrahydrate, cd no3 2.4h2o, acmc-20aljb, ksc174c1h, cadmium nitrate-water 1/4, cadmiumnitratetetrahydrate, cadmium ii nitrate tetrahydrate, nitric acid, cadmium salt tetrahydrate
PubChem CID 56924536
ChEBI CHEBI:86156
SMILES [Cd++].O[N+]([O-])=O.O[N+]([O-])=O

Specifications

Boiling Point 132°C
Melting Point 59°C
Color White
Physical Form Crystalline
Quantity 5 g
Assay Percent Range 99.999% (Metal basis)
Solubility Information Soluble in water,acetone,ethyl acetate,alcohol. Insoluble in nitric acid.
IUPAC Name cadmium(2+);dinitrate;tetrahydrate
Formula Weight 308.47 (236.41 Anhydrous)
Odor Weak
Appearance White crystalline
Density 2.45
Chemical Name or Material Cadmium nitrate tetrahydrate
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Cadmium nitrate tetrahydrate
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 2
  • Acute toxicity Category 3
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Carcinogenicity Category 1B
  • Germ cell mutagenicity Category 1B
  • Reproductive toxicity Category 1B
  • Specific target organ toxicity Category 1
Hazard Statement
  • H301-Toxic if swallowed.
  • H312-Harmful in contact with skin.
  • H330-Fatal if inhaled.
  • H340-May cause genetic defects.
  • H350-May cause cancer.
  • H360-May damage fertility or the unborn child.
  • H372-Causes damage to organs.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P202-Do not handle until all safety precautions have been read and understood.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contains : Cadmium(II) nitrate,tetrahydrate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.