missing translation for 'onlineSavingsMsg'
Learn More

Cadmium nitrate tetrahydrate, Puratronic™, 99.999% (metals basis), Thermo Scientific Chemicals

Catalog Number 11350889
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
10022-68-1
CdH2N2O6
238.44
MFCD00149626
IZIPRBSCGMISKX-UHFFFAOYSA-N
cadmium nitrate tetrahydrate, nitric acid, cadmium salt, tetrahydrate, cd no3 2.4h2o, acmc-20aljb, ksc174c1h, cadmium nitrate-water 1/4, cadmiumnitratetetrahydrate, cadmium ii nitrate tetrahydrate, nitric acid, cadmium salt tetrahydrate
56924536
CHEBI:86156
[Cd++].O[N+]([O-])=O.O[N+]([O-])=O
This item is not returnable. View return policy
10022-68-1
CdH2N2O6
238.44
MFCD00149626
IZIPRBSCGMISKX-UHFFFAOYSA-N
cadmium nitrate tetrahydrate, nitric acid, cadmium salt, tetrahydrate, cd no3 2.4h2o, acmc-20aljb, ksc174c1h, cadmium nitrate-water 1/4, cadmiumnitratetetrahydrate, cadmium ii nitrate tetrahydrate, nitric acid, cadmium salt tetrahydrate
56924536
CHEBI:86156
[Cd++].O[N+]([O-])=O.O[N+]([O-])=O

It is used for coloring glass and porcelain, and as a flash powder in photography. It is used as a laboratory reagent, in preparation of cadmium salts.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Boiling Point 132°C
Melting Point 59°C
Color White
Physical Form Crystalline
Quantity 25 g
Assay Percent Range 99.999% (Metal basis)
Solubility Information Soluble in water,acetone,ethyl acetate,alcohol. Insoluble in nitric acid.
IUPAC Name cadmium(2+);dinitrate;tetrahydrate
Formula Weight 308.47 (236.41 Anhydrous)
Odor Weak
Appearance White crystalline
Density 2.45 g/cm3
Chemical Name or Material Cadmium nitrate tetrahydrate
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'