missing translation for 'onlineSavingsMsg'
Learn More

Cadmium nitrate tetrahydrate, 99.9% (metals basis)

Product Code. 11358888
Change view
Click to view available options
Quantity:
50 g
250 g
Unit Size:
250g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11358888 250 g 250g
11368888 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11358888 Supplier Thermo Scientific Alfa Aesar Supplier No. 041732.30
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 10022-68-1 | CdH2N2O6 | 238.44 g/mol

Cadmium nitrate tetrahydrate is used in Photographic emulsions, coloring glass and porcelain, laboratory reagent, and the production of other cadmium compounds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10022-68-1
Molecular Formula CdH2N2O6
Molecular Weight (g/mol) 238.44
MDL Number MFCD00149626
InChI Key IZIPRBSCGMISKX-UHFFFAOYSA-N
Synonym cadmium nitrate tetrahydrate, nitric acid, cadmium salt, tetrahydrate, cd no3 2.4h2o, acmc-20aljb, ksc174c1h, cadmium nitrate-water 1/4, cadmiumnitratetetrahydrate, cadmium ii nitrate tetrahydrate, nitric acid, cadmium salt tetrahydrate
PubChem CID 56924536
ChEBI CHEBI:86156
IUPAC Name cadmium(2+);dinitrate;tetrahydrate
SMILES [Cd++].O[N+]([O-])=O.O[N+]([O-])=O

Specifications

Boiling Point 132°C
Melting Point 59°C
Color White
Physical Form Crystals
Quantity 250 g
Assay Percent Range 99.9% (Metals basis)
UN Number UN3087
Merck Index 14,1622
Solubility Information Soluble in water.(109g/100ml)at 0 C and (326g/100ml)at 60 C.Soluble in ammonia,acetone and alcohol.
Formula Weight 308.47 (236.41 Anhydrous)
Percent Purity 99.9%
Density 2.45
Assay (metals basis)
Chemical Name or Material Cadmium nitrate tetrahydrate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.