missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cadmium Isotopic Standard: Cd @ 10 μg/mL in 2% HNO3 (Enriched in Cd-106), VHG™
Description
Cadmium Isotopic Standard: Cd @ 10 μg/mL in 2% HNO3 (Enriched in Cd-106)
Specifications
Specifications
| Chemical Name or Material | Cadmium 106 |
| Methods | ICP-MS |
| Analyte or Component Names | Cadmium 106 |
| Shipping Condition | Room Temperature |
| Molecular Formula | 106Cd |
| Synonym | 106Cd |
| SMILES | [106Cd] |
| IUPAC Name | Cadmium 106 |
| Solvent or Matrix | Dilute HNO3 |
| Molecular Weight (g/mol) | 105.91 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?