missing translation for 'onlineSavingsMsg'
Learn More

Cadmium iodide, ultra dry, 99.9985% (metals basis)

Product Code. 11343857
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11343857 5 g 5g
11353857 1 g 1g
11333857 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11343857 Supplier Thermo Scientific Alfa Aesar Supplier No. 035683.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 7790-80-9 | CdI2 | 366.223 g/mol

Cadmium iodide is used in lithography, photography, electroplating and the manufacturing of phosphors.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Ampouled under argon
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7790-80-9
Molecular Formula CdI2
Molecular Weight (g/mol) 366.223
MDL Number MFCD00010920
InChI Key OKIIEJOIXGHUKX-UHFFFAOYSA-L
Synonym cadmium iodide, cadmium diiodide, unii-2f2upu4kcw, cadmium 2+ diiodide, 2f2upu4kcw, acmc-20aljc, cadmium 2+ ion diiodide, cadmium iodide trace metals basis 25g
PubChem CID 24635
IUPAC Name cadmium(2+);diiodide
SMILES [Cd+2].[I-].[I-]

Specifications

Boiling Point 787°C
Melting Point 387°C to 389°C
Color White to Yellow
Physical Form Beads
Quantity 5 g
UN Number UN2570
Merck Index 14,1621
Solubility Information It is soluble in water,ethanol,acetone,ether and ammonia.
Formula Weight 366.21
Percent Purity 99.9985%
Odor Odorless
Packaging Ampule
Sensitivity Hygroscopic; Light sensitive
Density 5.67 g/mL
Assay (metals basis)
Chemical Name or Material Cadmium iodide, Ultra dry
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.