missing translation for 'onlineSavingsMsg'
Learn More

Cadmium iodide, 98%

Product Code. 11439226
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11439226 100 g 100g
11429226 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11439226 Supplier Thermo Scientific Alfa Aesar Supplier No. A18079.22
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 7790-80-9 | CdI2 | 366.223 g/mol

It finds it use in photography, process engraving, lithography, analytical chemistry, and electroplating; Also used as in lubricants, phosphors. Used for cadmium electrode position and in photoconductors.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It finds it use in photography, process engraving, lithography, analytical chemistry, and electroplating; Also used as in lubricants, phosphors. Used for cadmium electrode position and in photoconductors.

Solubility
Very soluble in water, slightly soluble in acetone, ethanol and diethyl ether.

Notes
Light Sensitive & Hygroscopic. Store away from oxidizing agents, light and water/moisture. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store under inert gas.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7790-80-9
Molecular Formula CdI2
Molecular Weight (g/mol) 366.223
MDL Number MFCD00010920
InChI Key OKIIEJOIXGHUKX-UHFFFAOYSA-L
Synonym cadmium iodide, cadmium diiodide, unii-2f2upu4kcw, cadmium 2+ diiodide, 2f2upu4kcw, acmc-20aljc, cadmium 2+ ion diiodide, cadmium iodide trace metals basis 25g
PubChem CID 24635
IUPAC Name cadmium(2+);diiodide
SMILES [Cd+2].[I-].[I-]

Specifications

Boiling Point 787°C
Melting Point 387°C to 389°C
Quantity 100 g
UN Number UN2570
Merck Index 14,1621
Solubility Information Very soluble in water,slightly soluble in acetone,ethanol and diethyl ether.
Formula Weight 366.21
Percent Purity 98%
Odor Odorless
Sensitivity Hygroscopic; Light sensitive
Density 5.67
Chemical Name or Material Cadmium iodide
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.