missing translation for 'onlineSavingsMsg'
Learn More

Cadmium fluoride, Puratronic™, 99.99% (metals basis)

Product Code. 11359347
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11359347 100 g 100g
11349347 5 g 5g
11369347 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11359347 Supplier Thermo Scientific Alfa Aesar Supplier No. 040203.22
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 7790-79-6 | CdF2 | 150.41 g/mol

Cadmium fluoride is used in phosphors, glass and nuclear reactor controls. It is also used in high-temperature lubricants and to start crystals for lasers. It is used in oxygen-sensitive applications such as the production of metallic alloys and it is also used in limited medical treatment protocols. In addition, it can be used in synthetic organic chemistry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7790-79-6
Molecular Formula CdF2
Molecular Weight (g/mol) 150.41
MDL Number MFCD00010919
InChI Key LVEULQCPJDDSLD-UHFFFAOYSA-L
IUPAC Name cadmium(2+) difluoride
SMILES [F-].[F-].[Cd++]

Specifications

Boiling Point 1758°C
Melting Point 1100°C
Physical Form Powder
Quantity 100 g
UN Number UN2570
Merck Index 14,1619
Solubility Information Soluble in water (43.5g/L).
Formula Weight 150.4
Percent Purity 99.99%
Grade Puratronic
Odor Odorless
Density 6.64 g/mL
Assay (metals basis)
Chemical Name or Material Cadmium fluoride
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.