missing translation for 'onlineSavingsMsg'
Learn More

Cadmium fluoride, Puratronic™, 99.99% (metals basis), Thermo Scientific Chemicals

Catalog Number 11369347
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
7790-79-6
CdF2
150.41
MFCD00010919
LVEULQCPJDDSLD-UHFFFAOYSA-L
cadmium(2+) difluoride
[F-].[F-].[Cd++]
This item is not returnable. View return policy
7790-79-6
CdF2
150.41
MFCD00010919
LVEULQCPJDDSLD-UHFFFAOYSA-L
cadmium(2+) difluoride
[F-].[F-].[Cd++]

Cadmium fluoride is used in phosphors, glass and nuclear reactor controls. It is also used in high-temperature lubricants and to start crystals for lasers. It is used in oxygen-sensitive applications such as the production of metallic alloys and it is also used in limited medical treatment protocols. In addition, it can be used in synthetic organic chemistry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Boiling Point 1758°C
Melting Point 1100°C
Physical Form Powder
Quantity 25 g
UN Number UN2570
Merck Index 14,1619
Formula Weight 150.4
Percent Purity 99.99%
Grade Puratronic™
Odor Odorless
Density 6.64g/mL
Assay (metals basis)
Chemical Name or Material Cadmium fluoride
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'