missing translation for 'onlineSavingsMsg'
Learn More

Cadmium chloride, ultra dry, 99.998% (metals basis), Thermo Scientific Chemicals

Catalog Number 11386928
Click to view available options
Quantity:
5 g
25 g
10108-64-2
CdCl2
183.31
MFCD00010916
YKYOUMDCQGMQQO-UHFFFAOYSA-L
cadmium chloride, cadmium dichloride, caddy, vi-cad, kadmiumchlorid, caswell no. 135, ccris 114, hsdb 278, cadmium ii chloride
24947
CHEBI:35456
dichlorocadmium
[Cl-].[Cl-].[Cd++]
10108-64-2
CdCl2
183.31
MFCD00010916
YKYOUMDCQGMQQO-UHFFFAOYSA-L
cadmium chloride, cadmium dichloride, caddy, vi-cad, kadmiumchlorid, caswell no. 135, ccris 114, hsdb 278, cadmium ii chloride
24947
CHEBI:35456
dichlorocadmium
[Cl-].[Cl-].[Cd++]

Cadmium chloride is utilized in the preparation of CdTe nanocrystals, highly luminescent. It is also employed in analytical chemistry, photography, dyeing and calico printing, ingredient of electroplating baths, manufacture of special mirrors, the preparation of cadmium standard, analysis of sulfides, and testing pyridine bases.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Ampouled under argon
TRUSTED_SUSTAINABILITY
Boiling Point 960°C
Melting Point 568°C
Physical Form Beads
Quantity 5 g
UN Number UN2570
Merck Index 14,1617
Solubility Information Soluble in water and acetone.
Formula Weight 183.32
Percent Purity 99.998%
Odor Odorless
Packaging Ampule
Sensitivity Hygroscopic
Density 4.047 g/mL
Assay (metals basis)
Chemical Name or Material Cadmium chloride, Ultra dry
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'