missing translation for 'onlineSavingsMsg'
Learn More

Cadmium chloride, ultra dry, 99.998% (metals basis)

Product Code. 11376928
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11376928 25 g 25g
11386928 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11376928 Supplier Thermo Scientific Alfa Aesar Supplier No. 013667.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 10108-64-2 | CdCl2 | 183.31 g/mol

Cadmium chloride is utilized in the preparation of CdTe nanocrystals, highly luminescent. It is also employed in analytical chemistry, photography, dyeing and calico printing, ingredient of electroplating baths, manufacture of special mirrors, the preparation of cadmium standard, analysis of sulfides, and testing pyridine bases.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Ampouled under argon
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10108-64-2
Molecular Formula CdCl2
Molecular Weight (g/mol) 183.31
MDL Number MFCD00010916
InChI Key YKYOUMDCQGMQQO-UHFFFAOYSA-L
Synonym cadmium chloride, cadmium dichloride, caddy, vi-cad, kadmiumchlorid, caswell no. 135, ccris 114, hsdb 278, cadmium ii chloride
PubChem CID 24947
ChEBI CHEBI:35456
IUPAC Name dichlorocadmium
SMILES [Cl-].[Cl-].[Cd++]

Specifications

Boiling Point 960°C
Melting Point 568°C
Physical Form Beads
Quantity 25 g
UN Number UN2570
Merck Index 14,1617
Solubility Information Soluble in water and acetone.
Formula Weight 183.32
Percent Purity 99.998%
Odor Odorless
Packaging Ampule
Sensitivity Hygroscopic
Density 4.047 g/mL
Assay (metals basis)
Chemical Name or Material Cadmium chloride, Ultra dry
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.