missing translation for 'onlineSavingsMsg'
Learn More

Cadmium chloride hydrate, Puratronic™, 99.998% (metals basis)

Product Code. 11390879
Change view
Click to view available options
Quantity:
1 g
10 g
50 g
Unit Size:
10g
1g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11390879 1 g 1g
11370879 10 g 10g
11380879 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11390879 Supplier Thermo Scientific Alfa Aesar Supplier No. 010661.03
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 654054-66-7 | CdCl2 | 183.31 g/mol

Cadmium chloride hydrate is used in dyeing, printing, textiles and in metal finishing baths. It is also used as a reagent to make photographic paper and carbon paper. Further, it is utilized as an absorbing agent to absorb hydrogen sulfide as well as analytical reagent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 654054-66-7
Molecular Formula CdCl2
Molecular Weight (g/mol) 183.31
MDL Number MFCD00149627
InChI Key YKYOUMDCQGMQQO-UHFFFAOYSA-L
Synonym cadmium chloride, ultra dry, cdcl2.h2o, cadmium chloride hydrate, cadmium chloride 1-hydrate, cadmium chloride hydrate, puratronic, cadmium chloride monohydrate, puriss, cadmium chloride hydrate trace metals basis
PubChem CID 16211517
SMILES [Cl-].[Cl-].[Cd++]

Specifications

Physical Form Crystalline Powder
Quantity 1 g
UN Number UN2570
Merck Index 14,1617
IUPAC Name dichlorocadmium;hydrate
Formula Weight 183.32 (Anhydrous)
Percent Purity 99.998%
Grade Puratronic
Refractive Index 1.6513
Sensitivity Hygroscopic
Assay (metals basis)
Chemical Name or Material Cadmium chloride hydrate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.