missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cadmium chloride hydrate, Puratronic™, 99.998% (metals basis), Thermo Scientific Chemicals
28.00 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
1 g
10 g
50 g
Description
Cadmium chloride hydrate is used in dyeing, printing, textiles and in metal finishing baths. It is also used as a reagent to make photographic paper and carbon paper. Further, it is utilized as an absorbing agent to absorb hydrogen sulfide as well as analytical reagent.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Physical Form | Crystalline Powder |
Quantity | 10 g |
UN Number | UN2570 |
Merck Index | 14,1617 |
Solubility Information | Soluble in water and acetone. Slightly soluble in methanol. Insoluble in ether. |
IUPAC Name | cadmium(2+) dichloride |
Formula Weight | 183.32 (Anhydrous) |
Percent Purity | 99.998% |
Grade | Puratronic™ |
Refractive Index | 1.6513 |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'