missing translation for 'onlineSavingsMsg'
Learn More

Cadmium chloride hydrate, 99.99% (metals basis), Thermo Scientific Chemicals

Catalog Number 11343597
63.06 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.


Click to view available options
Quantity:
25 g
100 g
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
654054-66-7
CdCl2
183.31
MFCD00149627
YKYOUMDCQGMQQO-UHFFFAOYSA-L
cadmium chloride, ultra dry, cdcl2.h2o, cadmium chloride hydrate, cadmium chloride 1-hydrate, cadmium chloride hydrate, puratronic, cadmium chloride monohydrate, puriss, cadmium chloride hydrate trace metals basis
16211517
dichlorocadmium;hydrate
[Cl-].[Cl-].[Cd++]
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
654054-66-7
CdCl2
183.31
MFCD00149627
YKYOUMDCQGMQQO-UHFFFAOYSA-L
cadmium chloride, ultra dry, cdcl2.h2o, cadmium chloride hydrate, cadmium chloride 1-hydrate, cadmium chloride hydrate, puratronic, cadmium chloride monohydrate, puriss, cadmium chloride hydrate trace metals basis
16211517
dichlorocadmium;hydrate
[Cl-].[Cl-].[Cd++]

Cadmium chloride hydrate is used in laboratory chemicals and manufacture of substances.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Physical Form Powder
Quantity 25 g
UN Number UN2570
Merck Index 14,1617
Solubility Information Soluble in water.
Formula Weight 183.32 (Anhydrous)
Percent Purity 99.99%
Refractive Index 1.6513
Sensitivity Hygroscopic
Assay (metals basis)
Chemical Name or Material Cadmium chloride hydrate
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'