missing translation for 'onlineSavingsMsg'
Learn More

Cadmium chloride hemipentahydrate, 99+%

Product Code. 10318530 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
250 g
Packaging:
Glass bottle
Unit Size:
250g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10318530 250 g Glass bottle 250g
10308530 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10318530 Supplier Thermo Scientific Acros Supplier No. 197022500
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 7790-78-5 | 2·5 H2O | 228.34 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7790-78-5
Molecular Formula 2·5 H2O
Molecular Weight (g/mol) 228.34
MDL Number MFCD00799071

Specifications

Melting Point 34.0°C
Color White
Physical Form Crystalline Powder, Crystals and/or Chunks
Quantity 250 g
Assay Percent Range 79.5 to 81% (CdCl2) (Complexometry)
Linear Formula CdCl2·5/2H2O
Merck Index 15, 1622
Solubility Information Solubility in water: 1680g/L (20°C). Other solubilities: soluble in acetone, slightly soluble in ethanol and methanol
Formula Weight 228.34
Percent Purity ≥99%
Packaging Glass bottle
Chemical Name or Material Cadmium chloride hemipentahydrate, p.a.
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Cadmium chloride hemipentahydrate
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 2
  • Acute toxicity Category 3
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Carcinogenicity Category 1B
  • Germ cell mutagenicity Category 1B
  • Reproductive toxicity Category 1B
  • Specific target organ toxicity Category 1
Hazard Statement
  • H301-Toxic if swallowed.
  • H330-Fatal if inhaled.
  • H340-May cause genetic defects.
  • H350-May cause cancer.
  • H360FD-May damage fertility. May damage the unborn child.
  • H372-Causes damage to organs.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: Cadmium chloride hemipentahydrate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.