missing translation for 'onlineSavingsMsg'
Learn More

Cadmium chloride, anhydrous, ACS, 99.0% min, Thermo Scientific Chemicals

Catalog Number 11338068
41.66 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.


Click to view available options
Quantity:
100 g
500 g
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
10108-64-2
CdCl2
183.31
MFCD00010916
YKYOUMDCQGMQQO-UHFFFAOYSA-L
cadmium chloride, cadmium dichloride, caddy, vi-cad, kadmiumchlorid, caswell no. 135, ccris 114, hsdb 278, cadmium ii chloride
24947
CHEBI:35456
dichlorocadmium
[Cl-].[Cl-].[Cd++]
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
10108-64-2
CdCl2
183.31
MFCD00010916
YKYOUMDCQGMQQO-UHFFFAOYSA-L
cadmium chloride, cadmium dichloride, caddy, vi-cad, kadmiumchlorid, caswell no. 135, ccris 114, hsdb 278, cadmium ii chloride
24947
CHEBI:35456
dichlorocadmium
[Cl-].[Cl-].[Cd++]

Cadmium chloride is a precursor material for the preparation of cadmium sulfide. It is mainly utilized in dyeing, pigments, photocopying and electroplating of cadmium.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Preparation of cadmium sulfide, as analytical lab reagent, photography, manufacturing of special mirrors, ice-nucleating agent, lubricant, to absorb H2S in sulfide analysis
TRUSTED_SUSTAINABILITY
Boiling Point 960°C
Melting Point 568°C
Physical Form Powder
Quantity 100 g
UN Number UN2570
Merck Index 14,1617
Solubility Information Soluble in water,acetone; Slightly soluble in methanol; Insoluble in ether.
Formula Weight 183.32
Percent Purity ≤99.0%
Grade ACS
Odor Odorless
Sensitivity Hygroscopic
Density 4.047 g/mL
Chemical Name or Material Cadmium chloride, Anhydrous
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'