missing translation for 'onlineSavingsMsg'
Learn More

Cadmium chloride, 99%, for analysis, anhydrous, Thermo Scientific™

Click to view available options
Quantity:
100g
500g
Packaging:
Glass bottle
This item is not returnable. View return policy
10108-64-2
CdCl2
183.31
MFCD00010916
YKYOUMDCQGMQQO-UHFFFAOYSA-L
cadmium chloride, cadmium dichloride, caddy, vi-cad, kadmiumchlorid, caswell no. 135, ccris 114, hsdb 278, cadmium ii chloride
24947
CHEBI:35456
dichlorocadmium
[Cl-].[Cl-].[Cd++]
This item is not returnable. View return policy
10108-64-2
CdCl2
183.31
MFCD00010916
YKYOUMDCQGMQQO-UHFFFAOYSA-L
cadmium chloride, cadmium dichloride, caddy, vi-cad, kadmiumchlorid, caswell no. 135, ccris 114, hsdb 278, cadmium ii chloride
24947
CHEBI:35456
dichlorocadmium
[Cl-].[Cl-].[Cd++]

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Boiling Point 960.0°C
Color White
Melting Point 568.0°C
Quantity 100g
Assay Percent Range 98.5% min. (On dry substance) (Argentometry)
Linear Formula CdCl2
Fieser 04,485
Merck Index 15, 1622
Solubility Information Solubility in water: 1400g/L (20°C). Other solubilities: soluble in acetone,slightly soluble in methanol and ethanol,practically insoluble in ether
Formula Weight 183.31
Physical Form Powder
Percent Purity 99%
Grade Analysis
Insoluble Matter 0.01% max
Iron (Fe) 10ppm max.
Lead (Pb) 50ppm max.
Nitrate and Nitrite 0.003% max. (as NO3)
Ammonium 100ppm max.
Packaging Glass bottle
Solubility Solubility in water: 1400g/L (20°C). Other solubilities: soluble in acetone, slightly soluble in methanol and ethanol, practically insoluble in ether
Substances not ppt by H2S 0.3% max.
Sulfate 100ppm max.
Assay Conditions assay on dry substance
Copper (Cu) 10ppm max.
Chemical Name or Material Cadmium chloride, 99%
Show More Show Less

RUO – Research Use Only