missing translation for 'onlineSavingsMsg'
Learn More

Cadmium carbonate, >99% (metals basis), Thermo Scientific Chemicals

Product Code. p-4407294
Click to view available options
Quantity:
250 g
1 kg
Unit Size:
1kg
250g
This item is not returnable. View return policy

Product Code. 11371579

Brand: Thermo Scientific Alfa Aesar 012229.30

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Cadmium carbonate is the raw material for the production of pigments for ceramic and frits. Used as reagents.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 513-78-0
Molecular Formula CCdO3
Molecular Weight (g/mol) 172.42
MDL Number MFCD00010918
InChI Key GKDXQAKPHKQZSC-UHFFFAOYSA-L
Synonym cadmium carbonate, mikrokalcit, otavite, kalcit, supermikrokalcit, carbonic acid, cadmium salt 1:1, cadmium monocarbonate, chemcarb cdco3, caswell no. 134a, unii-616zvs3bly
PubChem CID 10564
IUPAC Name cadmium(2+);carbonate
SMILES [Cd++].[O-]C([O-])=O

Specifications

Melting Point 500°C (decomposition)
Color White
Physical Form Powder
Quantity 250 g
Assay Percent Range >99% (Metal basis)
Solubility Information Very slightly soluble in water. Soluble in dilute acids,concentrated solutions of ammonium salts.
Formula Weight 172.42
Odor Odorless
Appearance White powder
Density 4.26 g/cm3
Chemical Name or Material Cadmium carbonate
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.