missing translation for 'onlineSavingsMsg'
Learn More

Cadmium carbonate, 99.99% (metals basis), Thermo Scientific Chemicals

Product Code. 11331938
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11331938 100 g 100g
11341938 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11331938 Supplier Thermo Scientific Alfa Aesar Supplier No. 011864.22
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Cadmium carbonate, has been used as intermediates of manufacturing polyester, glass pigment flux, organic catalysts, plastic plasticizer, stabilizer,and, raw material for production of cadmium salt.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 513-78-0
Molecular Formula CCdO3
Molecular Weight (g/mol) 172.42
MDL Number MFCD00010918
InChI Key GKDXQAKPHKQZSC-UHFFFAOYSA-L
Synonym cadmium carbonate, mikrokalcit, otavite, kalcit, supermikrokalcit, carbonic acid, cadmium salt 1:1, cadmium monocarbonate, chemcarb cdco3, caswell no. 134a, unii-616zvs3bly
PubChem CID 10564
IUPAC Name cadmium(2+) carbonate
SMILES [Cd++].[O-]C([O-])=O

Specifications

Color White
Physical Form Lump/Powder
Quantity 100 g
Assay Percent Range 99.99%
Formula Weight 172.42
Odor Odorless
Appearance White powder/lumps
Chemical Name or Material Cadmium carbonate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.