missing translation for 'onlineSavingsMsg'
Learn More

Cadmium carbonate, 99.99% (metals basis), Thermo Scientific Chemicals

Catalog Number 11341938
Click to view available options
Quantity:
25 g
100 g
This item is not returnable. View return policy
513-78-0
CCdO3
172.42
MFCD00010918
GKDXQAKPHKQZSC-UHFFFAOYSA-L
cadmium carbonate, mikrokalcit, otavite, kalcit, supermikrokalcit, carbonic acid, cadmium salt 1:1, cadmium monocarbonate, chemcarb cdco3, caswell no. 134a, unii-616zvs3bly
10564
cadmium(2+) carbonate
[Cd++].[O-]C([O-])=O
This item is not returnable. View return policy
513-78-0
CCdO3
172.42
MFCD00010918
GKDXQAKPHKQZSC-UHFFFAOYSA-L
cadmium carbonate, mikrokalcit, otavite, kalcit, supermikrokalcit, carbonic acid, cadmium salt 1:1, cadmium monocarbonate, chemcarb cdco3, caswell no. 134a, unii-616zvs3bly
10564
cadmium(2+) carbonate
[Cd++].[O-]C([O-])=O

Cadmium carbonate, has been used as intermediates of manufacturing polyester, glass pigment flux, organic catalysts, plastic plasticizer, stabilizer,and, raw material for production of cadmium salt.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 500°C (decomposition)
Color White
Physical Form Lump/Powder
Quantity 25 g
Assay Percent Range 99.99%
Solubility Information It is insoluble in water and organic solvents,slightly soluble in aqueous acid,potassium cyanide,ammonium.
Formula Weight 172.42
Odor Odorless
Appearance White powder/lumps
Density 4.26 g/mL
Chemical Name or Material Cadmium carbonate
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'