missing translation for 'onlineSavingsMsg'
Learn More

Cadmium bromide, ultra dry, 99.999% (metals basis)

Product Code. 11323857
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11323857 5 g 5g
11303857 1 g 1g
11313857 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11323857 Supplier Thermo Scientific Alfa Aesar Supplier No. 035682.06
 Restricted Item
Estimated Shipment: 14-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 7789-42-6

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Ampouled under argon
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7789-42-6
MDL Number MFCD00015996
Synonym cadmium 2+ dibromide, cadmium 2+ ion dibromide, cadmium bromide, anhydrous 25g, cadmium bromide, anhydrous,

Specifications

Boiling Point 863°C
Melting Point 567°C
Color White to Yellow
Physical Form Beads
Quantity 5 g
UN Number UN2570
Merck Index 14,1615
Formula Weight 272.22
Percent Purity 99.999%
Odor Odorless
Packaging Ampule
Sensitivity Hygroscopic
Density 5.192 g/mL
Assay (metals basis)
Chemical Name or Material Cadmium bromide, Ultra dry
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.