missing translation for 'onlineSavingsMsg'
Learn More

Cadmium bromide, ultra dry, 99.999% (metals basis)

Product Code. p-7050129
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
This item is not returnable. View return policy

Product Code. 11303857

Brand: Thermo Scientific Alfa Aesar 035682.03

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 7789-42-6

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Ampouled under argon
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7789-42-6
MDL Number MFCD00015996
Synonym cadmium 2+ dibromide, cadmium 2+ ion dibromide, cadmium bromide, anhydrous 25g, cadmium bromide, anhydrous,

Specifications

Boiling Point 863°C
Melting Point 567°C
Color White to Yellow
Physical Form Beads
Quantity 1 g
UN Number UN2570
Merck Index 14,1615
Formula Weight 272.22
Percent Purity 99.999%
Odor Odorless
Packaging Ampule
Sensitivity Hygroscopic
Density 5.192g/mL
Assay (metals basis)
Chemical Name or Material Cadmium bromide, Ultra dry
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.