missing translation for 'onlineSavingsMsg'
Learn More

Cadmium bromide tetrahydrate, 98%, pure

Product Code. 15239896 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15239896 500 g 500g
15219896 25 g 25g
15229896 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15239896 Supplier Thermo Scientific Acros Supplier No. 453875000
 Restricted Item
Estimated Shipment: 05-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 13464-92-1 | Br2Cd·4H2O | 344.28 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13464-92-1
Molecular Formula Br2Cd·4H2O
Molecular Weight (g/mol) 344.28
InChI Key PANYFCXNZSEXSU-UHFFFAOYSA-L
Synonym cadmium dibromide, tetrahydrate, acmc-20alji, ksc497c4n, cadmium ii ; bromide tetrahydrate, cadmium 2+ dibromide tetrahydrate, cadmium 2+ ion tetrahydrate dibromide, cadmium bromide, tetrahydrate, reagent
PubChem CID 3084147
IUPAC Name cadmium(2+);dibromide;tetrahydrate
SMILES O.O.O.O.[Br-].[Br-].[Cd+2]

Specifications

Quantity 500 g
Assay Percent Range 97.5% min
Solubility Information Solubility in water: soluble.
Formula Weight 344.28
Percent Purity 98%
Grade Pure
Chemical Name or Material Cadmium bromide tetrahydrate
compliance-icons compliance-icons
Product Identifier
  • Cadmium bromide, tetrahydrate
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
Hazard Statement
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.